Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243516.1 | NP_001230445.1 | 5 - 6 | 723 | 60 | |
| XM_005671062.3 | XP_005671119.1 | 1065 | 60 | |||
| XM_013983315.2 | XP_013838769.1 | 699 | 60 | |||
| GenBank mRNA | AK344419.1 | - | 5 - 6 | 723 | 60 | |
| CB476425.1 | - | 3 - 4 | 289 | 60 | ||
More Information
Set Membership: |
|

AP3M1
Genomic Map