Assay Details
| Gene Symbol: DAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243563.1 | NP_001230492.1 | 9 - 10 | 914 | 77 | |
| XM_021093314.1 | XP_020948973.1 | 945 | 77 | |||
| XM_021093345.1 | XP_020949004.1 | 1292 | 77 | |||
| XM_021093392.1 | XP_020949051.1 | 942 | 77 | |||
| GenBank mRNA | AK348859.1 | - | 9 - 10 | 914 | 77 | |
| AK348984.1 | - | 8 - 9 | 884 | 77 | ||
More Information
Set Membership: |
|

DAPK2
Genomic Map