Assay Details
| Gene Symbol: LRP2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243566.2 | NP_001230495.2 | 8 - 9 | 1095 | 77 | |
| XM_021074532.1 | XP_020930191.1 | 1451 | 77 | |||
| XM_021074533.1 | XP_020930192.1 | 1938 | 77 | |||
| XM_021074534.1 | XP_020930193.1 | 1446 | 77 | |||
| XM_021074535.1 | XP_020930194.1 | 1738 | 77 | |||
| XM_021074536.1 | XP_020930195.1 | 1646 | 77 | |||
| XM_021074537.1 | XP_020930196.1 | 1126 | 77 | |||
| XM_021074538.1 | XP_020930197.1 | 1574 | 77 | |||
| XM_021074539.1 | XP_020930198.1 | 1394 | 77 | |||
| XM_021074540.1 | XP_020930199.1 | 1359 | 77 | |||
| GenBank mRNA | AK234106.1 | - | 8 - 9 | 1095 | 77 | |
More Information
Set Membership: |
|

LRP2BP
Genomic Map