Assay Details
| Gene Symbol: FH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243588.1 | NP_001230517.1 | 8 - 9 | 1240 | 69 | |
| XM_021064041.1 | XP_020919700.1 | 1275 | 69 | |||
| XM_021064042.1 | XP_020919701.1 | 1428 | 69 | |||
| XM_021064043.1 | XP_020919702.1 | 1386 | 69 | |||
| XM_021064044.1 | XP_020919703.1 | 1544 | 69 | |||
| GenBank mRNA | AK233410.1 | - | 8 - 9 | 1260 | 69 | |
| AK392470.1 | - | 8 - 9 | 1257 | 69 | ||
| AK398748.1 | - | 8 - 9 | 1266 | 69 | ||
| AY609689.1 | - | 8 - 9 | 1240 | 69 | ||
More Information
Set Membership: |
|

FH
Genomic Map