Assay Details
| Gene Symbol: AMZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243604.1 | NP_001230533.1 | 3 - 4 | 229 | 97 | |
| XM_005656929.3 | XP_005656986.1 | 202 | 97 | |||
| XM_013989897.2 | XP_013845351.1 | 163 | 97 | |||
| XM_021066162.1 | XP_020921821.1 | 378 | 97 | |||
| GenBank mRNA | AK238125.1 | - | 2 - 3 | 476 | 97 | |
| AY609674.1 | - | 3 - 4 | 229 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243604.1, XM_005656929.3, XM_013989897.2, XM_021066162.1 transcripts.
Set Membership: |
|

AMZ2
Genomic Map