Assay Details
| Gene Symbol: AMZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243604.1 | NP_001230533.1 | 8 - 9 | 1155 | 101 | |
| XM_005656927.3 | XP_005656984.1 | 1163 | 101 | |||
| XM_005656928.3 | XP_005656985.1 | 1136 | 101 | |||
| XM_005656929.3 | XP_005656986.1 | 1128 | 101 | |||
| XM_005656930.3 | XP_005656987.1 | 1127 | 101 | |||
| XM_005656931.3 | XP_005656988.1 | 1087 | 101 | |||
| XM_013989897.2 | XP_013845351.1 | 1089 | 101 | |||
| XM_021066162.1 | XP_020921821.1 | 1304 | 101 | |||
| XM_021066163.1 | XP_020921822.1 | 874 | 101 | |||
| XM_021066164.1 | XP_020921823.1 | 781 | 101 | |||
| GenBank mRNA | AK238125.1 | - | 7 - 8 | 1402 | 101 | |
| AY609674.1 | - | 8 - 9 | 1155 | 101 | ||
More Information
Set Membership: |
|

AMZ2
Genomic Map