Assay Details
| Gene Symbol: AP2M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243644.1 | NP_001230573.1 | 3 - 4 | 513 | 87 | |
| XM_005670021.3 | XP_005670078.1 | 513 | 87 | |||
| XM_021068490.1 | XP_020924149.1 | 378 | 87 | |||
| GenBank mRNA | AK235010.1 | - | 3 - 4 | 513 | 87 | |
| AK235168.1 | - | 2 - 3 | 365 | 87 | ||
| AK236737.1 | - | 3 - 4 | 491 | 87 | ||
| AK389446.1 | - | 3 - 4 | 453 | 87 | ||
| AK391250.1 | - | 2 - 3 | 444 | 87 | ||
| AK391363.1 | - | 3 - 4 | 443 | 87 | ||
| AK394991.1 | - | 3 - 4 | 551 | 87 | ||
| AK396047.1 | - | 3 - 4 | 485 | 87 | ||
| AK397798.1 | - | 3 - 4 | 489 | 87 | ||
| AK398326.1 | - | 3 - 4 | 476 | 87 | ||
| AK398964.1 | - | 3 - 4 | 489 | 87 | ||
More Information
Set Membership: |
|

AP2M1
Genomic Map