Assay Details
| Gene Symbol: RPP38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243673.1 | NP_001230602.1 | 2 - 3 | 284 | 91 | |
| XM_005668110.3 | XP_005668167.1 | 358 | 91 | |||
| GenBank mRNA | AK234545.1 | - | 2 - 3 | 280 | 91 | |
| AK238884.1 | - | 2 - 3 | 284 | 91 | ||
| AK399033.1 | - | 2 - 3 | 334 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243673.1, XM_005668110.3 transcripts.
Set Membership: |
|

RPP38
Genomic Map