Assay Details
| Gene Symbol: SNAPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243678.1 | NP_001230607.1 | 1 - 2 | 123 | 88 | |
| XM_005659861.1 | XP_005659918.1 | 164 | 88 | |||
| XM_013993311.2 | XP_013848765.1 | 164 | 88 | |||
| XM_021102402.1 | XP_020958061.1 | 164 | 88 | |||
| GenBank mRNA | AK345157.1 | - | 1 - 2 | 123 | 88 | |
| AK345748.1 | - | 1 - 2 | 89 | 88 | ||
More Information
Set Membership: |
|

SNAPC5
Genomic Map