Assay Details
| Gene Symbol: TMED5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243695.1 | NP_001230624.1 | 2 - 3 | 439 | 88 | |
| GenBank mRNA | AY610000.1 | - | 2 - 3 | 439 | 88 | |
More Information
Set Membership: |
|

TMED5
Genomic Map