Assay Details
| Gene Symbol: ADI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243698.1 | NP_001230627.1 | 1 - 2 | 175 | 89 | |
| GenBank mRNA | AK240417.1 | - | 1 - 2 | 174 | 89 | |
| AK350993.1 | - | 1 - 2 | 179 | 89 | ||
More Information
Set Membership: |
|

ADI1
Genomic Map