Assay Details
| Gene Symbol: MORN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243709.1 | NP_001230638.1 | 3 - 4 | 530 | 85 | |
| GenBank mRNA | AK351364.1 | - | 3 - 4 | 530 | 85 | |
More Information
Set Membership: |
|

MORN3
Genomic Map