Assay Details
| Gene Symbol: PPP1R14D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243710.1 | NP_001230639.1 | 1 - 2 | 350 | 78 | |
| XM_021062777.1 | XP_020918436.1 | 402 | 78 | |||
| XM_021062855.1 | XP_020918514.1 | 401 | 78 | |||
| XM_021063003.1 | XP_020918662.1 | 1209 | 78 | |||
| GenBank mRNA | AY610133.1 | - | 1 - 2 | 350 | 78 | |
More Information
Set Membership: |
|

PPP1R14D
Genomic Map