Assay Details
| Gene Symbol: RBMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244096.1 | NP_001231025.1 | 8 - 9 | 1006 | 101 | |
| XM_013986455.2 | XP_013841909.1 | 1061 | 101 | |||
| XM_021079411.1 | XP_020935070.1 | 1078 | 101 | |||
| XM_021079412.1 | XP_020935071.1 | 1061 | 101 | |||
| GenBank mRNA | AK235135.1 | - | 8 - 9 | 1006 | 101 | |
| AK236383.1 | - | 8 - 9 | 1025 | 101 | ||
| AK236451.1 | - | 8 - 9 | 1009 | 101 | ||
| AK236585.1 | - | 8 - 9 | 1010 | 101 | ||
| AK394814.1 | - | 8 - 9 | 1006 | 101 | ||
| AK397739.1 | - | 8 - 9 | 1061 | 101 | ||
More Information
Set Membership: |
|

RBMX
Genomic Map