Assay Details
| Gene Symbol: RPS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244106.1 | NP_001231035.1 | 5 - 6 | 852 | 85 | |
| XM_005665896.3 | XP_005665953.2 | 604 | 85 | |||
| GenBank mRNA | AK231549.1 | - | 5 - 6 | 499 | 85 | |
| AK240249.1 | - | 5 - 6 | 781 | 85 | ||
| AK349815.1 | - | 5 - 6 | 500 | 85 | ||
| AK349976.1 | - | 5 - 6 | 500 | 85 | ||
| AK350605.1 | - | 5 - 6 | 785 | 85 | ||
| AK397539.1 | - | 5 - 6 | 499 | 85 | ||
More Information
Set Membership: |
|

RPS10
Genomic Map