Assay Details
| Gene Symbol: HMGN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244112.1 | NP_001231041.1 | 1 - 2 | 217 | 81 | |
| NM_001244114.1 | NP_001231043.1 | 1 - 2 | 109 | 81 | ||
| GenBank mRNA | AK236192.1 | - | 1 - 2 | 200 | 81 | |
| AK236998.1 | - | 1 - 2 | 135 | 81 | ||
| AK347742.1 | - | 1 - 2 | 147 | 81 | ||
| AK349259.1 | - | 1 - 2 | 207 | 81 | ||
| AK350129.1 | - | 1 - 2 | 415 | 81 | ||
| AK391733.1 | - | 1 - 2 | 114 | 81 | ||
| AK397701.1 | - | 1 - 2 | 207 | 81 | ||
| BW968439.1 | - | 1 - 2 | 217 | 81 | ||
| EW418018.2 | - | 1 - 2 | 109 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244112.1, NM_001244114.1 transcripts.
Set Membership: |
|

HMGN2
Genomic Map