Assay Details
| Gene Symbol: CXCL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244128.1 | NP_001231057.1 | 3 - 4 | 524 | 100 | |
| GenBank mRNA | AK231165.1 | - | 3 - 4 | 562 | 100 | |
| AK231389.1 | - | 3 - 4 | 554 | 100 | ||
| AK231559.1 | - | 3 - 4 | 524 | 100 | ||
| AK233754.1 | - | 3 - 4 | 486 | 100 | ||
| AK233879.1 | - | 3 - 4 | 489 | 100 | ||
| AK234044.1 | - | 3 - 4 | 492 | 100 | ||
| AK345106.1 | - | 3 - 4 | 486 | 100 | ||
| AK345135.1 | - | 3 - 4 | 489 | 100 | ||
| AK345275.1 | - | 3 - 4 | 489 | 100 | ||
| AK346702.1 | - | 3 - 4 | 493 | 100 | ||
More Information
Set Membership: |
|

CXCL14
Genomic Map