Assay Details
| Gene Symbol: SCARB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244155.1 | NP_001231084.1 | 6 - 7 | 1149 | 70 | |
| XM_005666776.2 | XP_005666833.1 | 1144 | 70 | |||
| GenBank mRNA | AK395497.1 | - | 6 - 7 | 1149 | 70 | |
| AY609583.1 | - | 6 - 7 | 948 | 70 | ||
More Information
Set Membership: |
|

SCARB2
Genomic Map