Assay Details
| Gene Symbol: DHRS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244160.1 | NP_001231089.1 | 3 - 4 | 738 | 66 | |
| GenBank mRNA | AK231851.1 | - | 3 - 4 | 451 | 66 | |
| AK232729.1 | - | 3 - 4 | 510 | 66 | ||
| AK239997.1 | - | 3 - 4 | 492 | 66 | ||
| AY609687.1 | - | 3 - 4 | 738 | 66 | ||
More Information
Set Membership: |
|

DHRS7
Genomic Map