Assay Details
| Gene Symbol: PRIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244164.1 | NP_001231093.1 | 4 - 5 | 419 | 62 | |
| XM_021098097.1 | XP_020953756.1 | 491 | 62 | |||
| XM_021098098.1 | XP_020953757.1 | 473 | 62 | |||
| GenBank mRNA | AK351565.1 | - | 4 - 5 | 419 | 62 | |
| AK395443.1 | - | 4 - 5 | 421 | 62 | ||
More Information
Set Membership: |
|

PRIM2
Genomic Map