Assay Details
| Gene Symbol: LAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244255.1 | NP_001231184.1 | 3 - 4 | 539 | 75 | |
| NM_001244256.1 | NP_001231185.1 | 3 - 4 | 507 | 75 | ||
| GenBank mRNA | AK230639.1 | - | 3 - 4 | 539 | 75 | |
| AK230773.1 | - | 3 - 4 | 510 | 75 | ||
| AK231061.1 | - | 3 - 4 | 514 | 75 | ||
| AK235422.1 | - | 3 - 4 | 510 | 75 | ||
| AK343695.1 | - | 3 - 4 | 539 | 75 | ||
| AK392411.1 | - | 3 - 4 | 514 | 75 | ||
| AK398951.1 | - | 1 - 2 | 76 | |||
More Information
Set Membership: |
|

LAMP2
Genomic Map