Assay Details
| Gene Symbol: IPO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244301.1 | NP_001231230.1 | 2 - 3 | 188 | 64 | |
| GenBank mRNA | AK231050.1 | - | 2 - 3 | 188 | 64 | |
More Information
Set Membership: |
|

IPO5
Genomic Map