Assay Details
| Gene Symbol: SHFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244321.1 | NP_001231250.1 | 3 - 4 | 506 | 89 | |
| GenBank mRNA | AK351200.1 | - | 3 - 4 | 354 | 89 | |
| AK351221.1 | - | 3 - 4 | 506 | 89 | ||
More Information
Set Membership: |
|

SHFL
Genomic Map