Assay Details
| Gene Symbol: SDCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244414.2 | NP_001231343.2 | 2 - 3 | 132 | 73 | |
| XM_021088484.1 | XP_020944143.1 | 97 | 73 | |||
| XM_021088485.1 | XP_020944144.1 | 882 | 73 | |||
| XM_021088486.1 | XP_020944145.1 | 243 | 73 | |||
| XM_021088487.1 | XP_020944146.1 | 205 | 73 | |||
| XM_021088488.1 | XP_020944147.1 | 214 | 73 | |||
| XM_021088490.1 | XP_020944149.1 | 214 | 73 | |||
| GenBank mRNA | AK230648.1 | - | 2 - 3 | 100 | 73 | |
| AK230913.1 | - | 2 - 3 | 117 | 73 | ||
| AK230930.1 | - | 2 - 3 | 115 | 73 | ||
| AK230937.1 | - | 2 - 3 | 119 | 73 | ||
| AK230963.1 | - | 2 - 3 | 132 | 73 | ||
| AK343772.1 | - | 2 - 3 | 132 | 73 | ||
| AK347640.1 | - | 8 - 9 | 882 | 73 | ||
| AK389623.1 | - | 2 - 3 | 117 | 73 | ||
| AK389775.1 | - | 2 - 3 | 100 | 73 | ||
| AK389788.1 | - | 2 - 3 | 128 | 73 | ||
| AK389799.1 | - | 2 - 3 | 132 | 73 | ||
| AK389825.1 | - | 2 - 3 | 248 | 73 | ||
| AK399467.1 | - | 2 - 3 | 100 | 73 | ||
| AY609570.1 | - | 2 - 3 | 165 | 73 | ||
| CV866899.1 | - | 2 - 3 | 177 | 73 | ||
More Information
Set Membership: |
|

SDCBP
Genomic Map