Assay Details
| Gene Symbol: RHOV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244417.1 | NP_001231346.1 | 2 - 3 | 342 | 60 | |
| GenBank mRNA | AK239933.1 | - | 2 - 3 | 342 | 60 | |
More Information
Set Membership: |
|

RHOV
Genomic Map