Assay Details
| Gene Symbol: TCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244436.1 | NP_001231365.1 | 6 - 7 | 1081 | 102 | |
| XM_005670816.3 | XP_005670873.1 | 1078 | 102 | |||
| XM_005670817.3 | XP_005670874.1 | 1034 | 102 | |||
| XM_005670818.3 | XP_005670875.1 | 1045 | 102 | |||
| XM_005670819.3 | XP_005670876.1 | 863 | 102 | |||
| XM_005670820.3 | XP_005670877.1 | 1175 | 102 | |||
| XM_005670821.3 | XP_005670878.1 | 1137 | 102 | |||
| XM_021071885.1 | XP_020927544.1 | 1375 | 102 | |||
| GenBank mRNA | AK234715.1 | - | 6 - 7 | 1024 | 102 | |
| AK240444.1 | - | 6 - 7 | 1030 | 102 | ||
| AK398652.1 | - | 6 - 7 | 1030 | 102 | ||
| AK398863.1 | - | 6 - 7 | 1021 | 102 | ||
More Information
Set Membership: |
|

TCN2
Genomic Map