Assay Details
| Gene Symbol: PRDX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244474.1 | NP_001231403.1 | 3 - 4 | 373 | 116 | |
| GenBank mRNA | AK349436.1 | - | 3 - 4 | 373 | 116 | |
| AK390553.1 | - | 3 - 4 | 322 | 116 | ||
| AK392270.1 | - | 3 - 4 | 341 | 116 | ||
| AK400020.1 | - | 3 - 4 | 373 | 116 | ||
| AY610343.1 | - | 3 - 4 | 309 | 116 | ||
| GQ913659.1 | - | 3 - 4 | 327 | 116 | ||
More Information
Set Membership: |
|

PRDX2
Genomic Map