Assay Details
| Gene Symbol: ICMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244481.1 | NP_001231410.1 | 4 - 5 | 722 | 61 | |
| GenBank mRNA | CF793853.1 | - | 1 - 2 | 99 | 61 | |
| CN161844.1 | - | 4 - 5 | 687 | 61 | ||
More Information
Set Membership: |
|

ICMT
Genomic Map