Assay Details
| Gene Symbol: SHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244513.1 | NP_001231442.1 | 2 - 3 | 560 | 80 | |
| GenBank mRNA | BX918823.1 | - | 2 - 3 | 339 | 80 | |
More Information
Set Membership: |
|

SHH
Genomic Map