Assay Details
| Gene Symbol: PLAAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244514.1 | NP_001231443.1 | 1 - 2 | 76 | 60 | |
| GenBank mRNA | AK346775.1 | - | 1 - 2 | 54 | 60 | |
| CJ011354.1 | - | 1 - 2 | 76 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244514.1 transcripts.
Set Membership: |
|

PLAAT3
Genomic Map