Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244530.1 | NP_001231459.1 | 2 - 3 | 430 | 77 | |
| XM_005664650.2 | XP_005664707.1 | 404 | 77 | |||
| XM_021093375.1 | XP_020949034.1 | 366 | 77 | |||
| GenBank mRNA | AK240174.1 | - | 2 - 3 | 311 | 77 | |
| AK240588.1 | - | 2 - 3 | 430 | 77 | ||
| AK344904.1 | - | 2 - 3 | 181 | 77 | ||
More Information
Set Membership: |
|

EMP3
Genomic Map