Assay Details
| Gene Symbol: SFT2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244534.1 | NP_001231463.1 | 7 - 8 | 522 | 73 | |
| GenBank mRNA | AK234386.1 | - | 7 - 8 | 510 | 73 | |
| AK392532.1 | - | 7 - 8 | 499 | 73 | ||
| AK400085.1 | - | 7 - 8 | 525 | 73 | ||
| EW622209.2 | - | 7 - 8 | 535 | 73 | ||
More Information
Set Membership: |
|

SFT2D1
Genomic Map