Assay Details
| Gene Symbol: TMEM106A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244554.1 | NP_001231483.1 | 4 - 5 | 478 | 89 | |
| XM_005656948.3 | XP_005657005.1 | 412 | 89 | |||
| GenBank mRNA | AK230947.1 | - | 4 - 5 | 478 | 89 | |
| AK391495.1 | - | 4 - 5 | 478 | 89 | ||
More Information
Set Membership: |
|

TMEM106A
Genomic Map