Assay Details
| Gene Symbol: CLN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244565.1 | NP_001231494.1 | 2 - 3 | 859 | 108 | |
| XM_013995178.2 | XP_013850632.2 | 851 | 108 | |||
| XM_013995218.2 | XP_013850672.2 | 769 | 108 | |||
| XM_021074631.1 | XP_020930290.1 | 772 | 108 | |||
| XM_021074632.1 | XP_020930291.1 | 655 | 108 | |||
| XM_021074633.1 | XP_020930292.1 | 741 | 108 | |||
| XM_021074634.1 | XP_020930293.1 | 720 | 108 | |||
| XM_021074635.1 | XP_020930294.1 | 772 | 108 | |||
| XM_021074636.1 | XP_020930295.1 | 772 | 108 | |||
| XM_021074637.1 | XP_020930296.1 | 772 | 108 | |||
| XM_021074639.1 | XP_020930298.1 | 772 | 108 | |||
| XM_021074640.1 | XP_020930299.1 | 772 | 108 | |||
| XM_021074641.1 | XP_020930300.1 | 772 | 108 | |||
| XM_021074642.1 | XP_020930301.1 | 772 | 108 | |||
| GenBank mRNA | AK234438.1 | - | 2 - 3 | 859 | 108 | |
| AK390192.1 | - | 3 - 4 | 1043 | 108 | ||
| AK392579.1 | - | 2 - 3 | 835 | 108 | ||
More Information
Set Membership: |
|

CLN8
Genomic Map