Assay Details
| Gene Symbol: BATF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244571.1 | NP_001231500.1 | 1 - 2 | 270 | 83 | |
| GenBank mRNA | AK346632.1 | - | 1 - 2 | 270 | 83 | |
More Information
Set Membership: |
|

BATF
Genomic Map