Assay Details
| Gene Symbol: BATF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244571.1 | NP_001231500.1 | 2 - 3 | 380 | 65 | |
| GenBank mRNA | AK346632.1 | - | 2 - 3 | 380 | 65 | |
More Information
Set Membership: |
|

BATF
Genomic Map