Assay Details
| Gene Symbol: NDUFA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244646.1 | NP_001231575.1 | 1 - 2 | 200 | 68 | |
| GenBank mRNA | AK344861.1 | - | 1 - 2 | 118 | 68 | |
| AK400086.1 | - | 1 - 2 | 130 | 68 | ||
| CK466967.1 | - | 1 - 2 | 86 | 68 | ||
More Information
Set Membership: |
|

NDUFA13
Genomic Map