Assay Details
| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244667.1 | NP_001231596.1 | 3 - 4 | 337 | 93 | |
| NM_001244668.1 | NP_001231597.1 | 2 - 3 | 291 | 93 | ||
| XM_021091068.1 | XP_020946727.1 | 2895 | 93 | |||
| GenBank mRNA | AK232270.1 | - | 3 - 4 | 331 | 93 | |
| AK348469.1 | - | 1 - 2 | 135 | 93 | ||
| AK395209.1 | - | 3 - 4 | 330 | 93 | ||
| DT331776.1 | - | 3 - 4 | 326 | 93 | ||
| FD632152.1 | - | 2 - 3 | 285 | 93 | ||
| FS709250.1 | - | 2 - 3 | 285 | 93 | ||
| FS712898.1 | - | 3 - 4 | 290 | 93 | ||
More Information
Set Membership: |
|

DNAL4
Genomic Map