Assay Details
| Gene Symbol: RPL36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244687.1 | NP_001231616.1 | 3 - 4 | 260 | 55 | |
| GenBank mRNA | AK231197.1 | - | 2 - 3 | 444 | 55 | |
| AK348810.1 | - | 3 - 4 | 258 | 55 | ||
| AK350306.1 | - | 3 - 4 | 260 | 55 | ||
| AK351192.1 | - | 3 - 4 | 259 | 55 | ||
| AK392191.1 | - | 3 - 4 | 260 | 55 | ||
More Information
Set Membership: |
|

RPL36
Genomic Map