Assay Details
| Gene Symbol: RUBCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244715.1 | NP_001231644.1 | 5 - 6 | 745 | 58 | |
| XM_005670134.3 | XP_005670191.1 | 776 | 58 | |||
| XM_005670135.3 | XP_005670192.1 | 775 | 58 | |||
| XM_013982359.2 | XP_013837813.1 | 776 | 58 | |||
| XM_021068219.1 | XP_020923878.1 | 776 | 58 | |||
| XM_021068220.1 | XP_020923879.1 | 1056 | 58 | |||
| XM_021068221.1 | XP_020923880.1 | 776 | 58 | |||
| XM_021068222.1 | XP_020923881.1 | 1056 | 58 | |||
| XM_021068223.1 | XP_020923882.1 | 253 | 58 | |||
| XM_021068224.1 | XP_020923883.1 | 776 | 58 | |||
More Information
Set Membership: |
|

RUBCN
Genomic Map