Assay Details
| Gene Symbol: JAML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244730.1 | NP_001231659.1 | 5 - 6 | 649 | 93 | |
| XM_005653697.3 | XP_005653754.1 | 714 | 93 | |||
| XM_005653698.3 | XP_005653755.1 | 2876 | 93 | |||
| XM_005653699.3 | XP_005653756.1 | 705 | 93 | |||
| XM_005653701.3 | XP_005653758.1 | 744 | 93 | |||
| XM_013977422.2 | XP_013832876.1 | 1188 | 93 | |||
| XM_013977439.2 | XP_013832893.1 | 784 | 93 | |||
| XM_013977441.1 | XP_013832895.1 | 794 | 93 | |||
| XM_021102035.1 | XP_020957694.1 | 1251 | 93 | |||
| XM_021102036.1 | XP_020957695.1 | 1446 | 93 | |||
| GenBank mRNA | AK238962.1 | - | 5 - 6 | 649 | 93 | |
| AK389867.1 | - | 5 - 6 | 93 | |||
| AK398411.1 | - | 3 - 4 | 1027 | 93 | ||
More Information
Set Membership: |
|

JAML
Genomic Map