Assay Details
| Gene Symbol: CBR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244732.1 | NP_001231661.1 | 2 - 3 | 544 | 101 | |
| GenBank mRNA | AK347045.1 | - | 2 - 3 | 544 | 101 | |
More Information
Set Membership: |
|

CBR3
Genomic Map