Assay Details
| Gene Symbol: NUBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244801.1 | NP_001231730.1 | 4 - 5 | 526 | 61 | |
| XM_005655129.3 | XP_005655186.1 | 638 | 61 | |||
| XM_013987749.2 | XP_013843203.1 | 668 | 61 | |||
| GenBank mRNA | AK239101.1 | - | 4 - 5 | 526 | 61 | |
More Information
Set Membership: |
|

NUBP2
Genomic Map