Assay Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244811.1 | NP_001231740.1 | 4 - 5 | 696 | 82 | |
| GenBank mRNA | AK235771.1 | - | 4 - 5 | 696 | 82 | |
More Information
Set Membership: |
|

LRRC57
Genomic Map