Assay Details
| Gene Symbol: TREML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243504.1 | NP_001230433.1 | 1 - 2 | 99 | 102 | |
| XM_021098040.1 | XP_020953699.1 | 83 | 102 | |||
| GenBank mRNA | AK236921.1 | - | 1 - 2 | 99 | 102 | |
| AK236955.1 | - | 1 - 2 | 88 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243504.1, XM_021098040.1 transcripts.
Set Membership: |
|

TREML1
Genomic Map