Assay Details
| Gene Symbol: SQOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244238.1 | NP_001231167.1 | 2 - 3 | 95 | 123 | |
| GenBank mRNA | AY609727.1 | - | 2 - 3 | 95 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001244238.1 transcripts.
Set Membership: |
|

SQOR
Genomic Map