Assay Details
| Gene Symbol: GMFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244461.1 | NP_001231390.1 | 7 | 1320 | 118 | |
| XM_021078752.1 | XP_020934411.1 | 1383 | 118 | |||
| GenBank mRNA | AK344488.1 | - | 7 | 1320 | 118 | |
More Information
Set Membership: |
|

GMFB
Genomic Map