Assay Details
| Gene Symbol: LOC102160148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097573.1 | XP_020953232.1 | 42 | 72 | ||
| Gene Symbol: LOC100515837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK345052.1 | - | 1 - 2 | 155 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021097573.1 transcripts.
Set Membership: |
|

LOC102160148
Genomic Map