Assay Details
| Gene Symbol: TAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013992499.2 | XP_013847953.2 | 3102 | 119 | ||
| XM_021086945.1 | XP_020942604.1 | 2504 | 119 | |||
| XM_021086951.1 | XP_020942610.1 | 2308 | 119 | |||
| XM_021086959.1 | XP_020942618.1 | 2458 | 119 | |||
| XM_021086965.1 | XP_020942624.1 | 2266 | 119 | |||
| XM_021086968.1 | XP_020942627.1 | 2362 | 119 | |||
More Information
Set Membership: |
|

TAB2
Genomic Map