Assay Details
| Gene Symbol: MIR19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031536.2 | 1 | 44 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-18a | MIR18A |
MIR18, MIRN18, MIRN18A, ssc-mir-18, ssc-mir-18a |
-57.0 | Sense | Chr. 11 - 60734791 - 60734882 on NCBI Build Sscrofa11.1 |
| Downstream | microRNA mir-20a | MIR20A |
MIR20, MIRN20, MIRN20A, ssc-mir-20, ssc-mir-20a |
89.0 | Sense | Chr. 11 - 60735109 - 60735179 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR19A
Genomic Map